Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methylformamide 95%

Supplier: AMBEED, INC
Danger

A614798-1G A614798-250MG A614798-5G
76992-168EA 69 USD InStock
76992-168 76992-170 76992-172
N-Methoxy-N-methylformamide 95%
N-Methoxy-N-methylformamide
Precaución: for Research Use Only


Formula: C₃H₇NO₂
MDL Number: MFCD11226240
CAS Number: 32117-82-1
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty