Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5R,6S)-5,6-Diphenyl-2-morpholinone 97%

Supplier: AMBEED, INC
Warning

A615988-1G A615988-250MG A615988-25G A615988-5G
77005-474EA 17 USD InStock
77005-474 77005-476 77005-478 77005-480
(5R,6S)-5,6-Diphenyl-2-morpholinone 97%
(5R,6S)-5,6-Diphenyl-2-morpholinone
Precaución: for Research Use Only


Formula: C₁₆H₁₅NO₂
MDL Number: MFCD09751026
CAS Number: 282735-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty