Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-amino-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate 98%

Supplier: AMBEED, INC
Warning

A617764-100MG A617764-250MG
76992-186EA 35 USD InStock
76992-186 76992-188
tert-Butyl 3-amino-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate 98%
tert-Butyl 3-amino-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₈N₄O₂
MDL Number: MFCD12964106
CAS Number: 398491-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty