Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2,6-dioxo-8-(thiophen-2-yl)hexahydro-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborol-4-ium-8-uide 95%

Supplier: AMBEED, INC
Warning

A619606-10G A619606-1G
77018-108EA 54 USD InStock
77018-108 77018-110
4-Methyl-2,6-dioxo-8-(thiophen-2-yl)hexahydro-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborol-4-ium-8-uide 95%
4-Methyl-2,6-dioxo-8-(thiophen-2-yl)hexahydro-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborol-4-ium-8-uide
Precaución: for Research Use Only


Formula: C₉H₁₀BNO₄S
MW: 239.05 g/mol
MDL Number: MFCD15144786
CAS Number: 1158984-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty