Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 4-oxocyclopentane-1,2-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A619756-100MG A619756-1G A619756-250MG A619756-5G
77035-244EA 8 USD InStock
77035-244 77035-246 77035-248 77035-250
Dimethyl 4-oxocyclopentane-1,2-dicarboxylate 97%
Dimethyl 4-oxocyclopentane-1,2-dicarboxylate
Precaución: for Research Use Only


Formula: C₉H₁₂O₅
MW: 200.18 g/mol
MDL Number: MFCD00154556
CAS Number: 6453-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty