Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A623140-100MG A623140-1G A623140-250MG A623140-5G
76992-212EA 34 USD InStock
76992-212 76992-214 76992-216 76992-218
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Precaución: for Research Use Only


Formula: C₅₀H₅₇O₄P
MW: 752.959 g/mol
MDL Number: MFCD10567009
CAS Number: 791616-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty