Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pazopanib hydrochloride 99+%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A623193-10MG A623193-1MG A623193-100MG A623193-250MG A623193-5G
77018-132EA 7 USD InStock
77018-132 77018-134 77684-501 77684-502 77684-503
Pazopanib hydrochloride 99+%, Ambeed.Inc
Pazopanib hydrochloride
Precaución: For research use only.


Formula: C₂₁H₂₃N₇O₂S·HCl
MW: 452.5·36.5 g/mol
MDL Number: MFCD12546138
CAS Number: 635702-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty