Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indolin-7-ol 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A623913-1G A623913-250MG A623913-100MG
77018-136EA 465 USD InStock
77018-136 77018-138 77684-504
Indolin-7-ol 95%, Ambeed.Inc
Indolin-7-ol
Precaución: For research use only.


Formula: C₈H₉NO
MDL Number: MFCD11617214
CAS Number: 4770-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty