Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-benzoylbenzoate 98%

Supplier: AMBEED, INC
Warning

A624223-100G A624223-25G A624223-500G
77035-294EA 35 USD InStock
77035-294 77035-296 77035-298
Methyl-2-benzoylbenzoate 98%
Methyl 2-benzoylbenzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₂O₃
MW: 240.2540 g/mol
MDL Number: MFCD00017187
CAS Number: 606-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty