Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-iminodiacetic acid 98%

Supplier: AMBEED, INC
Warning

A624740-100G A624740-25G A624740-5G
77005-530EA 147 USD InStock
77005-530 77005-532 77005-534
Fmoc-iminodiacetic acid 98%
Fmoc-iminodiacetic acid
Precaución: for Research Use Only


Formula: C₁₉H₁₇NO₆
MDL Number: MFCD01321017
CAS Number: 112918-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty