Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-L-alanine 98%

Supplier: AMBEED, INC
Warning

A625328-100MG A625328-1G A625328-1MG
77005-536EA 16 USD InStock
77005-536 77005-538 77005-540
3-Chloro-L-alanine 98%
3-Chloro-L-alanine
Precaución: for Research Use Only


Formula: C₃H₆ClNO₂
MDL Number: MFCD00046536
CAS Number: 2731-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty