Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxythiophene 95%

Supplier: AMBEED, INC
Danger

A625542-100MG A625542-1G A625542-250MG
77035-316EA 45 USD InStock
77035-316 77035-318 77035-320
3-Hydroxythiophene 95%
3-Hydroxythiophene
Precaución: for Research Use Only


Formula: C₄H₄OS
MDL Number: MFCD12026375
CAS Number: 17236-59-8
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty