Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Pyridyl)-2-propyn-1-ol 98+%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A629119-1G A629119-250MG A629119-5G A629119-100MG
77018-174EA 59 USD InStock
77018-174 77018-176 77018-178 77684-513
3-(2-Pyridyl)-2-propyn-1-ol 98+%, Ambeed.Inc
3-(2-Pyridyl)-2-propyn-1-ol
Precaución: For research use only.


Formula: C₈H₇NO
MDL Number: MFCD00168870
CAS Number: 29768-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty