Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Naphthol AS-E phosphate (KG-501) 97%

Supplier: AMBEED, INC
Warning

A631283-10MG A631283-1MG A631283-50MG A631283-5MG
77035-340EA 35 USD InStock
77035-340 77035-342 77035-344 77035-346
Naphthol AS-E phosphate (KG-501) 97%
Naphthol AS-E phosphate (KG-501)
Precaución: for Research Use Only


Formula: C₁₇H₁₃ClNO₅P
CAS Number: 18228-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty