Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl isobutyroylmalonate 97%

Supplier: AMBEED, INC
Warning

A631836-1G A631836-250MG A631836-5G
77005-608EA 38 USD InStock
77005-608 77005-610 77005-612
Diethyl isobutyroylmalonate 97%
Diethyl isobutyroylmalonate
Precaución: for Research Use Only


Formula: C₁₁H₁₈O₅
MW: 230.26 g/mol
MDL Number: MFCD00048557
CAS Number: 21633-78-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty