Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-(pyrimidin-2-yl)ethanone hydrobromide 95%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A632943-1G A632943-5G
77018-230EA 132 USD InStock
77018-230 77684-518
2-Bromo-1-(pyrimidin-2-yl)ethanone hydrobromide 95%, Ambeed.Inc
2-Bromo-1-(pyrimidin-2-yl)ethanone hydrobromide
Precaución: For research use only.


Formula: C₆H₆Br₂N₂O
MDL Number: MFCD27920889
CAS Number: 1588441-02-4
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty