Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A633620-250MG A633620-5G A633620-1G
77018-232EA 19 USD InStock
77018-232 77018-234 77684-519
6-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%, Ambeed.Inc
6-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: For research use only.


Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD08059288
CAS Number: 174648-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty