Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,7-Dichloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A638756-100MG A638756-1G A638756-250MG A638756-5G
77018-260EA 34 USD InStock
77018-260 77018-262 77018-264 77018-266
5,7-Dichloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
5,7-Dichloro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₀Cl₃N
MW: 238.54 g/mol
MDL Number: MFCD05861557
CAS Number: 73075-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty