Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butylazetidin-3-ol 95%

Supplier: AMBEED, INC
Danger

A639032-10G A639032-1G A639032-250MG A639032-25G A639032-5G
77005-646EA 18 USD InStock
77005-646 77005-648 77005-650 77005-652 77005-654
1-tert-Butylazetidin-3-ol 95%
1-tert-Butylazetidin-3-ol
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD02323245
CAS Number: 13156-04-2
UN: 1325
ADR: 4.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty