Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-2-fluoro-6-methoxyphenyl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A639543-1G A639543-250MG A639543-5G
77018-278EA 60 USD InStock
77018-278 77018-280 77018-282
(3-Bromo-2-fluoro-6-methoxyphenyl)boronic acid 98%
(3-Bromo-2-fluoro-6-methoxyphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₇BBrFO₃
MDL Number: MFCD12026725
CAS Number: 1309980-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty