Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-N-methylpropanamide 95%

Supplier: AMBEED, INC
Warning

A640768-100G A640768-1G A640768-25G A640768-5G
77005-666EA 1350 USD InStock
77005-666 77005-668 77005-670 77005-672
3-Hydroxy-N-methylpropanamide 95%
3-Hydroxy-N-methylpropanamide
Precaución: for Research Use Only


Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD13188569
CAS Number: 6830-81-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty