Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1H-imidazol-1-ylacetate 95%

Supplier: AMBEED, INC
Warning

A643400-10G A643400-1G A643400-250MG A643400-5G
77005-698EA 113 USD InStock
77005-698 77005-700 77005-702 77005-704
Ethyl-1H-imidazol-1-ylacetate 95%
Ethyl-1H-imidazol-1-ylacetate
Precaución: for Research Use Only


Formula: C₇H₁₀N₂O₂
MDL Number: MFCD05149485
CAS Number: 17450-34-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty