Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluorobenzo[c][1,2]oxaborol-1(3H)-ol 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A647056-100MG A647056-250MG A647056-1G
77018-316EA 19 USD InStock
77018-316 77018-318 77684-530
6-Fluorobenzo[c][1,2]oxaborol-1(3H)-ol 95%, Ambeed.Inc
6-Fluorobenzo[c][1,2]oxaborol-1(3H)-ol
Precaución: For research use only.


Formula: C₇H₆BFO₂
MDL Number: MFCD10699481
CAS Number: 174671-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty