Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methyl-5-nitroindoline-2,3-dione 98%

Supplier: AMBEED, INC
Warning

A647274-1G A647274-250MG
76992-370EA 46 USD InStock
76992-370 76992-372
7-Methyl-5-nitroindoline-2,3-dione 98%
7-Methyl-5-nitroindoline-2,3-dione
Precaución: for Research Use Only


Formula: C₉H₆N₂O₄
MDL Number: MFCD00448204
CAS Number: 70343-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty