Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(methoxymethyl)fluorobenzene 95%

Supplier: AMBEED, INC
Warning

A647574-100MG A647574-1G A647574-250MG A647574-5G
76992-374EA 15 USD InStock
76992-374 76992-376 76992-378 76992-380
2-Bromo-4-(methoxymethyl)fluorobenzene 95%
2-Bromo-4-(methoxymethyl)fluorobenzene
Precaución: for Research Use Only


Formula: C₈H₈BrFO
MW: 219.051 g/mol
MDL Number: MFCD04116324
CAS Number: 887268-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty