Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Acetamido-8-nitro-3,4-Dihydro-2H-1,5-benzodioxepine 97%

Supplier: AMBEED, INC
Warning

A652223-100MG A652223-1G A652223-250MG
77035-470EA 50 USD InStock
77035-470 77035-472 77035-474
7-Acetamido-8-nitro-3,4-Dihydro-2H-1,5-benzodioxepine 97%
7-Acetamido-8-nitro-3,4-Dihydro-2H-1,5-benzodioxepine
Precaución: for Research Use Only


Formula: C₁₁H₁₂N₂O₅
MDL Number: MFCD00508726
CAS Number: 81864-61-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty