Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-α-4-dibromophenylacetate 98%

Supplier: AMBEED, INC
Danger

A659728-1G A659728-250MG A659728-5G
76992-456EA 40 USD InStock
76992-456 76992-458 76992-460
Ethyl-α-4-dibromophenylacetate 98%
Ethyl-alpha-4-dibromophenylacetate
Precaución: for Research Use Only


Formula: C₁₀H₁₀Br₂O₂
MDL Number: MFCD09953497
CAS Number: 77143-76-1
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty