Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine 98%

Supplier: AMBEED, INC
Warning

A659963-100MG A659963-1G A659963-250MG A659963-5G A659963-25G
77005-732EA 37 USD InStock
77005-732 77005-734 77005-736 77005-738 77005-740 77673-796
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine
Precaución: for Research Use Only


Formula: C₂₄H₁₆F₅NO₄
MW: 477.39 g/mol
MDL Number: MFCD00270612
CAS Number: 205526-32-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty