Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(p-Tolyl)-2-naphthylamine 98+%

Supplier: AMBEED, INC
Warning

A661746-1G A661746-250MG A661746-5G
77018-406EA 56 USD InStock
77018-406 77018-408 77018-410
N-(p-Tolyl)-2-naphthylamine 98+%
N-(p-Tolyl)-2-naphthylamine
Precaución: for Research Use Only


Formula: C₁₇H₁₅N
MDL Number: MFCD00222912
CAS Number: 644-16-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty