Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Boc-methylamino)-3-pyridineboronic acid 98%

Supplier: AMBEED, INC
Warning

A662837-100MG A662837-1G A662837-250MG A662837-5G
76992-464EA 36 USD InStock
76992-464 76992-466 76992-468 76992-470
6-(Boc-methylamino)-3-pyridineboronic acid 98%
6-(Boc-methylamino)-3-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇BN₂O₄
MDL Number: MFCD12756422
CAS Number: 1218790-80-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty