Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene 96%

Supplier: AMBEED, INC
Warning

A663954-100G A663954-1G A663954-25G A663954-5G
77035-550EA 127 USD InStock
77035-550 77035-552 77035-554 77035-556
(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene 96%
(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene
Precaución: for Research Use Only


Formula: C₁₃H₁₆O₂
MDL Number: MFCD09263486
CAS Number: 110567-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty