Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate 95%

Supplier: AMBEED, INC
Danger

A668039-100MG A668039-1G A668039-250MG A668039-5G
77018-438EA 12 USD InStock
77018-438 77018-440 77018-442 77018-444
(R)-tert-Butyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate 95%
(R)-tert-Butyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₅S
MDL Number: MFCD06809586
CAS Number: 127423-61-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty