Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethyl 4-boronobenzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A668301-1G A668301-250MG A668301-5G
77018-446EA 166 USD InStock
77018-446 77018-448 77018-450
N,N-Diethyl 4-boronobenzenesulphonamide 98%
N,N-Diethyl 4-boronobenzenesulphonamide
Precaución: for Research Use Only


Formula: C₈H₁₂BNO₄S
MW: 257.11 g/mol
MDL Number: MFCD06659846
CAS Number: 850568-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty