Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl ditelluride 98%

Supplier: AMBEED, INC
Warning

A671477-100MG A671477-1G A671477-250MG A671477-5G
77005-796EA 9 USD InStock
77005-796 77005-798 77005-800 77005-802
Diphenyl ditelluride 98%
Diphenyl ditelluride
Precaución: for Research Use Only


Formula: C₁₂H₁₀Te₂
MDL Number: MFCD00192106
CAS Number: 32294-60-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty