Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate 95%

Supplier: AMBEED, INC
Warning

A671599-1G A671599-250MG A671599-5G
77035-612EA 39 USD InStock
77035-612 77035-614 77035-616
2,5-Dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate 95%
2,5-Dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₀N₂O₆
MDL Number: MFCD01318593
CAS Number: 64191-06-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty