Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TGR5-Receptor-Agonist 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A672110-10MG A672110-1MG A672110-25MG A672110-5MG A672110-50MG
77018-470EA 35 USD InStock
77018-470 77018-472 77018-474 77018-476 77684-579
TGR5-Receptor-Agonist 98%, Ambeed.Inc
TGR5-Receptor-Agonist
Precaución: For research use only.


Formula: C₁₈H₁₄Cl₂N₂O₂
MDL Number: MFCD17169972
CAS Number: 1197300-24-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty