Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-7-nitrobenzo[c][1,2,5]oxadiazole 95%

Supplier: AMBEED, INC
Danger

A674697-1G A674697-250MG
76992-540EA 410 USD InStock
76992-540 76992-542
4-Bromo-7-nitrobenzo[c][1,2,5]oxadiazole 95%
4-Bromo-7-nitrobenzo[c][1,2,5]oxadiazole
Precaución: for Research Use Only


Formula: C₆H₂BrN₃O₃
MDL Number: MFCD26383600
CAS Number: 35128-56-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty