Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-phenoxyaniline 95%

Supplier: AMBEED, INC
Warning

A675067-100MG A675067-1G A675067-250MG
77005-816EA 32 USD InStock
77005-816 77005-818 77005-820
2-Methyl-4-phenoxyaniline 95%
2-Methyl-4-phenoxyaniline
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
MDL Number: MFCD08688317
CAS Number: 13024-16-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty