Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydrazinobenzenesulfonamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A675626-1000G A675626-25G A675626-500G
77005-822EA 131 USD InStock
77005-822 77005-824 77005-826 77005-828 77005-830
4-Hydrazinobenzenesulfonamide hydrochloride 97%
4-Hydrazinobenzenesulfonamide hydrochloride
Precaución: for Research Use Only


Formula: C₆H₉N₃O₂S
MDL Number: MFCD00052262
CAS Number: 17852-52-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty