Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Morpholin-4-ylacetohydrazide 95%

Supplier: AMBEED, INC
Warning

A678795-1G A678795-250MG A678795-5G
77035-654EA 59 USD InStock
77035-654 77035-656 77035-658
2-Morpholin-4-ylacetohydrazide 95%
2-Morpholin-4-ylacetohydrazide
Precaución: for Research Use Only


Formula: C₆H₁₃N₃O₂
MDL Number: MFCD02269932
CAS Number: 770-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty