Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hydroxyethylferrocene 98%

Supplier: AMBEED, INC
Warning

A682274-100G A682274-250MG A682274-25G A682274-5G
77018-570EA 236 USD InStock
77018-570 77018-572 77018-574 77018-576 77018-578
1-Hydroxyethylferrocene 98%
1-Hydroxyethylferrocene
Precaución: for Research Use Only


Formula: C₁₂H₅FeO
MDL Number: MFCD00013752
CAS Number: 1277-49-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty