Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-1-bromo-2-chlorobenzene 95%

Supplier: AMBEED, INC
Warning

A685761-250MG A685761-5G
76992-618EA 19 USD InStock
76992-618 76992-620
4-(Benzyloxy)-1-bromo-2-chlorobenzene 95%
4-(Benzyloxy)-1-bromo-2-chlorobenzene
Precaución: for Research Use Only


Formula: C₁₃H₁₀BrClO
MW: 297.57 g/mol
MDL Number: MFCD16251708
CAS Number: 729590-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty