Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hexyloxybenzoic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A686374-1G A686374-25G A686374-5G A686374-100G
77018-620EA 5 USD InStock
77018-620 77018-622 77018-624 77684-594
4-Hexyloxybenzoic acid 95%, Ambeed.Inc
4-Hexyloxybenzoic acid
Precaución: For research use only.


Formula: C₁₃H₁₈O₃
MDL Number: MFCD00013991
CAS Number: 1142-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty