Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-4-iodoanisole 95%

Supplier: AMBEED, INC
Warning

A686387-1G A686387-250MG A686387-25G A686387-5G
76992-622EA 27 USD InStock
76992-622 76992-624 76992-626 76992-628
2,6-Difluoro-4-iodoanisole 95%
2,6-Difluoro-4-iodoanisole
Precaución: for Research Use Only


Formula: C₇H₅F₂IO
MDL Number: MFCD07368787
CAS Number: 886762-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty