Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Iodonicotinic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A693349-5G A693349-1G
76992-666EA 21 USD InStock
76992-666 77684-600
6-Iodonicotinic acid 95%, Ambeed.Inc
6-Iodonicotinic acid
Precaución: For research use only.


Formula: C₆H₄INO₂
MDL Number: MFCD03426924
CAS Number: 13054-02-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty