Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methylbenzyl bromide 95%

Supplier: AMBEED, INC
Danger

A698441-100MG A698441-1G A698441-250MG A698441-5G
77035-762EA 9 USD InStock
77035-762 77035-764 77035-766 77669-886
4-Bromo-3-methylbenzyl bromide 95%
4-Bromo-3-methylbenzyl bromide
Precaución: for Research Use Only


Formula: C₈H₈Br₂
MW: 263.957 g/mol
MDL Number: MFCD09909629
CAS Number: 27561-51-9
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty