Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorocinnamic aldehyde 97%

Supplier: AMBEED, INC
Warning

A698788-1G A698788-250MG
77005-962EA 173 USD InStock
77005-962 77005-964
2,6-Difluorocinnamic aldehyde 97%
2,6-Difluorocinnamic aldehyde
Precaución: for Research Use Only


Formula: C₉H₆F₂O
MDL Number: MFCD07782064
CAS Number: 117338-43-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty