Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-(trifluoromethyl)phenol 95%

Supplier: AMBEED, INC
Warning

A700725-100MG A700725-1G A700725-250MG
77018-734EA 29 USD InStock
77018-734 77018-736 77018-738
2-Amino-3-(trifluoromethyl)phenol 95%
2-Amino-3-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₆F₃NO
MDL Number: MFCD15526973
CAS Number: 106877-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty