Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3,5-dimethylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A701199-100MG A701199-1G A701199-5G A701199-250MG
77005-980EA 46 USD InStock
77005-980 77005-982 77005-984 77673-922
2-Hydroxy-3,5-dimethylbenzaldehyde 95%
2-Hydroxy-3,5-dimethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD05625710
CAS Number: 24623-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty