Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-4-propoxybenzoate 95%

Supplier: AMBEED, INC
Warning

A705340-1G A705340-5G
76992-718EA 110 USD InStock
76992-718 76992-720
Methyl 3-bromo-4-propoxybenzoate 95%
Methyl 3-bromo-4-propoxybenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₃
MDL Number: MFCD06801019
CAS Number: 216965-51-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty